
Valifenalate

Synonym(s):
N-[(1-Methylethoxy)carbonyl]-L-valyl-3-(4-chlorophenyl)-β-alanine methyl ester, Methyl N-(isopropoxycarbonyl)-L-valyl-(3RS)-3-(4-chlorophenyl)-β-alaninate
Empirical Formula (Hill Notation):
C19H27ClN2O5
CAS Number:
Molecular Weight:
398.88
Products
Shipping
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing