Skip to Content
Merck
Undecane analytical standard

Undecane

Undecane analytical standard
Synonym(s):
n-Undecane, Hendecane
Linear Formula:
CH3(CH2)9CH3
CAS Number:
Molecular Weight:
156.31
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing