Skip to Content
Merck
Tritolyl phosphate technical grade, mixture of isomers, 90%

Tritolyl phosphate

Tritolyl phosphate technical grade, mixture of isomers, 90%
Synonym(s):
Tricresyl phosphate, Tritolyl phosphate
Linear Formula:
(CH3C6H4O)3PO
CAS Number:
Molecular Weight:
368.36

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing