tri-Sodium phosphate dodecaydrate
Synonym(s):
Sodium phosphate tribasic dodecahydrate, TSP, Trisodium phosphate (tert) dodecahydrate, Trisodium phosphate dodecahydrate, tri-Sodium phosphate dodecahydrate
Linear Formula:
Na3PO4 · 12H2O
CAS Number:
Molecular Weight:
380.12
Products
Product Category
Brand
Formula Weight
Product Line
Showing 1-2 of 2
Compare
Product Number
Description
Pricing