Skip to Content
Merck
Triphenylphosphine diiodide technical grade, 90%

Triphenylphosphine diiodide

Triphenylphosphine diiodide technical grade, 90%
Synonym(s):
Diiodotriphenylphosphorane
Linear Formula:
(C6H5)3PI2
CAS Number:
Molecular Weight:
516.09

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing