trans-4,5-Dihydroxy-1,2-dithiane
Synonym(s):
trans-1,2-Dithiane-4,5-diol, Oxidized DTT
Empirical Formula (Hill Notation):
C4H8O2S2
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing