
trans-3-Hydroxycinnamic acid

Synonym(s):
(E)-3-(3-Hydroxyphenyl)acrylic acid, m-Coumaric acid
Linear Formula:
HOC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
164.16
Products
Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1