![Thiobencarb PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/397/485/1108d6d9-8dac-4086-bc92-878ea56660df/640/1108d6d9-8dac-4086-bc92-878ea56660df.png)
Thiobencarb
![Thiobencarb PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/397/485/1108d6d9-8dac-4086-bc92-878ea56660df/640/1108d6d9-8dac-4086-bc92-878ea56660df.png)
Empirical Formula (Hill Notation):
C12H16ClNOS
Products
Shipping
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing