
Tetrahydroberberine

Synonym(s):
(±)-Canadine, (±)-Tetrahydroberberine, 5,8,13,13a-tetrahydro-9,10-dimethoxy-6H-benzo[g]-1,3-benzodioxolo[5,6-alpha]quinolizine, Canadine
Empirical Formula (Hill Notation):
C20H21NO4
CAS Number:
Molecular Weight:
339.39
Products
Color
Feature
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing