(−)-Terpinen-4-ol
Synonym(s):
(R)-1-Isopropyl-4-methyl-3-cyclohexen-1-ol, (R)-p-Menth-1-en-4-ol
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Products
Shipping
Product Category
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing