Synperonic® F 108
Synonym(s):
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)
CAS Number:
Products
Shipping
Product Category
Brand
Melting Point (°C)
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing