![Swertiamarin analytical standard](/deepweb/assets/sigmaaldrich/product/structures/215/793/f8d89c97-28aa-4a1a-bb74-4f1730412906/640/f8d89c97-28aa-4a1a-bb74-4f1730412906.png)
Swertiamarin
![Swertiamarin analytical standard](/deepweb/assets/sigmaaldrich/product/structures/215/793/f8d89c97-28aa-4a1a-bb74-4f1730412906/640/f8d89c97-28aa-4a1a-bb74-4f1730412906.png)
Synonym(s):
(4aR,5R,6S)-5-Ethenyl-6-(β-D-glucopyranosyloxy)-4,4a,5,6-tetrahydro-4a-hydroxy1H,3H-pyrano[3,4-c]pyran-1-one, 4,4a,5,6-Tetrahydro-4aα-hydroxy-1-oxo-5β-vinyl-1H,3H-pyrano[3,4-c]pyran-6-yl β-D-glucopyranoside, Swertiamarine, Swertiamaroside
Empirical Formula (Hill Notation):
C16H22O10
CAS Number:
Molecular Weight:
374.34
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing