
Sumatriptan succinate Related Compound C

Synonym(s):
{3-[2-(Dimethylamino)ethyl]-1-(hydroxymethyl)-1H-indol-5-yl}-N-methylmethanesulfonamide hemisuccinate salt, [3-[2-(dimethylamino)ethyl]-1-(hydroxymethyl)-1H-indol-5-yl]-N-methylmethanesulfonamide succinate salt
Empirical Formula (Hill Notation):
C15H23N3O3S·0.5C4H6O4
Molecular Weight:
384.47
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2