Sulfamethoxazole impurity F
Synonym(s):
4-Amino-N-(3-methyl-5-isoxazolyl)benzenesulfonamide
Empirical Formula (Hill Notation):
C10H11N3O3S
CAS Number:
Molecular Weight:
253.28
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing