Sodium molybdate
Synonym(s):
Sodium molybdate dihydrate
Empirical Formula (Hill Notation):
MoNa2O4 · 2H2O
CAS Number:
Molecular Weight:
241.95
Products
Product Category
Brand
Formula Weight
Product Line
Showing 1-2 of 2
Compare
Product Number
Description
Pricing