Silybin B
Synonym(s):
(2R,3R)-3,5,7-Trihydroxy-2-[(2S,3S)-3-(4-hydroxy-3-methoxyphenyl)-2-hydroxymethyl-2,3-dihydrobenzo[1,4]dioxin-6-yl]-4-chromanone, (2S,3S)-2,3-Dihydro-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-6-[(2R,3R)-3,5,7-trihydroxy-4-oxobenzopyran-2-yl]benzodioxin
Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing