Skip to Content
Merck
sec-Butylmagnesium chloride solution 2.0 M in diethyl ether

sec-Butylmagnesium chloride solution

sec-Butylmagnesium chloride solution 2.0 M in diethyl ether
Linear Formula:
CH3CH2CH(CH3)MgCl
CAS Number:
Molecular Weight:
116.87

Products

Shipping
Product Category
Formula Weight
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing