![Saxitoxin dihydrochloride solution 20 μg/g in hydrochloric acid (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/182/112/67cd80e0-6e8f-4bf8-99b8-b0c5b0db0414/640/67cd80e0-6e8f-4bf8-99b8-b0c5b0db0414.png)
Saxitoxin dihydrochloride solution
![Saxitoxin dihydrochloride solution 20 μg/g in hydrochloric acid (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/182/112/67cd80e0-6e8f-4bf8-99b8-b0c5b0db0414/640/67cd80e0-6e8f-4bf8-99b8-b0c5b0db0414.png)
Synonym(s):
3aS,4R,10aS)-2,6-Diamino-4-{[(aminocarbonyl)oxy]methyl}-3a,4,8,9-tetrahydro-1H,10H-pyrrolo[1,2-c]purine-10,10-diol dihydrochloride, STX
Empirical Formula (Hill Notation):
C10H17N7O4 · 2HCl
CAS Number:
Molecular Weight:
372.21
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1