Skip to Content
Merck
[Sar9, Met(O2)11]-Substance P ≥95% (HPLC)

[Sar9, Met(O2)11]-Substance P

[Sar9, Met(O2)11]-Substance P ≥95% (HPLC)
Synonym(s):
Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Sar-Leu-Met[O2]-NH2
Empirical Formula (Hill Notation):
C64H100N18O15S
CAS Number:
Molecular Weight:
1393.66

Products

Product Category
Brand
Formula Weight
Gene Alias
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing