Skip to Content
Merck
Roscovitine ≥98% (TLC)

Roscovitine

Roscovitine ≥98% (TLC)
Synonym(s):
2-(R)-[[9-(1-Methylethyl)-6-[(phenylmethyl)amino]-9H-purin-2-yl]amino]-1-butanol, 6-(Benzylamino)-2(R)-[[1-(hydroxymethyl)propyl]amino]-9-isopropylpurine, Roscovitine
Empirical Formula (Hill Notation):
C19H26N6O
CAS Number:
Molecular Weight:
354.45

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing