
Ranitidine Impurity D

Synonym(s):
Demethylamino Ranitidine Acetamide Sodium
Empirical Formula (Hill Notation):
C12H18N3NaO4S
CAS Number:
Molecular Weight:
323.34
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing