Skip to Content
Merck
Rabeprazole Related Compound D certified reference material, pharmaceutical secondary standard

Rabeprazole Related Compound D

Rabeprazole Related Compound D certified reference material, pharmaceutical secondary standard
Synonym(s):
Rabeprazole sulfone, 2-{[4-(3-Methoxypropoxy)-3-methyl-2-pyridyl]methylsulfonyl}-1-H-benzimidazole
Empirical Formula (Hill Notation):
C18H21N3O4S
CAS Number:
Molecular Weight:
375.44

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade