Protriptyline hydrochloride
Synonym(s):
N-Methyl-5H-dibenzo[a,d]cycloheptene-5-propanamine hydrochloride
Empirical Formula (Hill Notation):
C19H21N · HCl
CAS Number:
Molecular Weight:
299.84
Products
Shipping
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing