Potassium diphenylphosphide solution
Synonym(s):
Diphenylphosphine potassium salt, KPPh2
Linear Formula:
(C6H5)2PK
CAS Number:
Molecular Weight:
224.28
Products
Brand
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing