![Plasminogen activator inhibitor 1 (PAI-1) human recombinant, expressed in E. coli, ≥90% (SDS-PAGE)](/deepweb/assets/sigmaaldrich/product/images/217/551/32463bc9-0114-4892-adcb-4bba1b0df12b/640/32463bc9-0114-4892-adcb-4bba1b0df12b.jpg)
Plasminogen activator inhibitor 1 (PAI-1)
![Plasminogen activator inhibitor 1 (PAI-1) human recombinant, expressed in E. coli, ≥90% (SDS-PAGE)](/deepweb/assets/sigmaaldrich/product/images/217/551/32463bc9-0114-4892-adcb-4bba1b0df12b/640/32463bc9-0114-4892-adcb-4bba1b0df12b.jpg)
Synonym(s):
PAIE, PLANH1, Plasminogen activator inhibitor, β-migrating endothelial cell-derived type, Serpin E1 PAI-1, Serpine 1
CAS Number:
Products
Product Category
Brand
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing