![Picoxystrobin PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/692/882/799b08f1-7f77-4ecb-a52b-ca2f86d65ce5/640/799b08f1-7f77-4ecb-a52b-ca2f86d65ce5.png)
Picoxystrobin
![Picoxystrobin PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/692/882/799b08f1-7f77-4ecb-a52b-ca2f86d65ce5/640/799b08f1-7f77-4ecb-a52b-ca2f86d65ce5.png)
Synonym(s):
Methyl (E)-α-methoxymethylene-2-(3-trifluoromethyl-2-pyridyloxymethyl)phenylacetate
Empirical Formula (Hill Notation):
C18H16F3NO4
CAS Number:
Molecular Weight:
367.32
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2