Skip to Content
Merck
Phenylephrine Related Compound E Pharmaceutical Secondary Standard; Certified Reference Material

Phenylephrine Related Compound E

Phenylephrine Related Compound E Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
1-(3-Hydroxyphenyl)-2-[methyl(phenylmethyl)amino]ethanone hydrochloride, 2-[Benzyl(methyl)amino]-1-(3-hydroxyphenyl)ethanone hydrochloride, Benzylphenylephrone hydrochloride
Empirical Formula (Hill Notation):
C16H17NO2 · HCl
CAS Number:
Molecular Weight:
291.77

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique