Skip to Content
Merck
PHA-543613 ≥98% (HPLC)

PHA-543613

PHA-543613 ≥98% (HPLC)
Synonym(s):
N-[(3R)-1-Azabicyclo[2.2.2]oct-3-yl]furo[2,3-c]pyridine-5-carboxamide
Empirical Formula (Hill Notation):
C15H17N3O2
CAS Number:
Molecular Weight:
271.31

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing