
Peonidin chloride

Synonym(s):
3,4′,5,7-Tetrahydroxy-3′-methoxyflavylium chloride, Paeonidin chloride, Peonidol chloride
Empirical Formula (Hill Notation):
C16H13ClO6
CAS Number:
Molecular Weight:
336.72
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1