![p-Divinylbenzene 85%](/deepweb/assets/sigmaaldrich/product/structures/275/778/8c14be1d-0eeb-465a-9ebe-f32b331c32fa/640/8c14be1d-0eeb-465a-9ebe-f32b331c32fa.png)
p-Divinylbenzene
![p-Divinylbenzene 85%](/deepweb/assets/sigmaaldrich/product/structures/275/778/8c14be1d-0eeb-465a-9ebe-f32b331c32fa/640/8c14be1d-0eeb-465a-9ebe-f32b331c32fa.png)
Empirical Formula (Hill Notation):
C10H10
CAS Number:
Molecular Weight:
130.19
Products
Brand
Formula Weight
Purity
Chemical Composition
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing