
PDAM

Synonym(s):
1-Pyrenyldiazomethane, Pyrene, 1-(diazomethyl)
Empirical Formula (Hill Notation):
C17H10N2
CAS Number:
Molecular Weight:
242.27
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing