Skip to Content
IN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
PCR segmented microplate
Products
Product Category
plates (10)
Color
Manufacturer
Wells
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
AXYPCR96SGA
PCR segmented microplate,
size 96 wells, assorted colors, pkg of 5x10plates/cs
Expand
View Pricing
AXYPCR96SGB
PCR segmented microplate,
size 96 wells, blue, pkg of 5x10plates/cs
Expand
View Pricing
AXYPCR96SGBK
PCR segmented microplate,
size 96 wells, black, pkg of 5x10plates/cs
Expand
View Pricing
AXYPCR96SGCS
PCR segmented microplate,
size 96 wells, clear, pkg of 5x10plates/cs, sterile
Expand
View Pricing
AXYPCR96SGG
PCR segmented microplate,
size 96 wells, green, pkg of 5x10plates/cs
Expand
View Pricing
AXYPCR96SGO
PCR segmented microplate,
size 96 wells, orange, pkg of 5x10plates/cs
Expand
View Pricing
AXYPCR96SGR
PCR segmented microplate,
size 96 wells, red, pkg of 5x10plates/cs
Expand
View Pricing
AXYPCR96SGV
PCR segmented microplate,
size 96 wells, violet, pkg of 5x10plates/cs
Expand
View Pricing
AXYPCR96SGW
PCR segmented microplate,
size 96 wells, white, pkg of 5x10plates/cs
Expand
View Pricing
AXYPCR96SGY
PCR segmented microplate,
size 96 wells, yellow, pkg of 5x10plates/cs
Expand
View Pricing