Skip to Content
Merck
Oxaprozin pharmaceutical secondary standard, certified reference material

Oxaprozin

Oxaprozin pharmaceutical secondary standard, certified reference material
Synonym(s):
4,5-Diphenyl-2-oxazolepropanoic acid, Daypro, Oxaprozin
Empirical Formula (Hill Notation):
C18H15NO3
CAS Number:
Molecular Weight:
293.32

Products

Shipping
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade