Skip to Content
Merck
Olmesartan Medoxomil Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material

Olmesartan Medoxomil Related Compound A

Olmesartan Medoxomil Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
3,6-Dihydro-6,6-dimethyl-2-propyl-3-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-4H-furo[3,4-d]imidazol-4-one, 1-{[2′-(1H-Tetrazol-5-yl)biphenyl-4-yl]methyl}-4,4-dimethyl-2-propyl-1H-furo[3,4-d]imidazol-6(4H)-one
Empirical Formula (Hill Notation):
C24H24N6O2
CAS Number:
Molecular Weight:
428.49

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade