Skip to Content
Merck
N,N-Diisopropylcarbamoyl chloride 98%

N,N-Diisopropylcarbamoyl chloride

N,N-Diisopropylcarbamoyl chloride 98%
Linear Formula:
[(CH3)2CH]2NCOCl
CAS Number:
Molecular Weight:
163.65
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing