Skip to Content
Merck
Nitric acid c(HNO) = 10 mol/l (10 N)

Nitric acid solution

Nitric acid c(HNO) = 10 mol/l (10 N)
Synonym(s):
Nitric acid solution
Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Reaction Type
Technique
Showing 1-2 of 2