![Nicergoline impurity D European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/137/210/bb64b4c6-c8bd-4edc-899d-5cbc5ae59dfe/640/bb64b4c6-c8bd-4edc-899d-5cbc5ae59dfe.png)
Nicergoline impurity D
![Nicergoline impurity D European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/137/210/bb64b4c6-c8bd-4edc-899d-5cbc5ae59dfe/640/bb64b4c6-c8bd-4edc-899d-5cbc5ae59dfe.png)
Synonym(s):
5-Bromopyridine-3-carboxylic acid, 5-Bromonicotinic acid
Empirical Formula (Hill Notation):
C6H4BrNO2
CAS Number:
Molecular Weight:
202.01
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing