Skip to Content
Merck
Nevadensin ≥90% (LC/MS-ELSD)

Nevadensin

Nevadensin ≥90% (LC/MS-ELSD)
Synonym(s):
5,7-Dihydroxy 4′,6,8-trimethoxyflavone, Pedunculin
Empirical Formula (Hill Notation):
C18H16O7
CAS Number:
Molecular Weight:
344.32

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing