Skip to Content
Merck
N-Acetyl-D-sphingosine ≥98.0% (TLC)

N-Acetyl-D-sphingosine

N-Acetyl-D-sphingosine ≥98.0% (TLC)
Synonym(s):
(2S,3R,4E)-2-(Acetylamino)-4-octadecene-1,3-diol, N-Acetoyl-D-erythro-sphingosine, Acetyl ceramide, C2 Ceramide (d18:1/2:0), C2 ceramide
Empirical Formula (Hill Notation):
C20H39NO3
Molecular Weight:
341.53

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing