N-[3-(Trimethoxysilyl)propyl]aniline
Synonym(s):
Trimethoxy[3-(phenylamino)propyl]silane, (3-Anilinopropyl)trimethoxysilane, N-[3-(Trimethoxysilyl)propyl]aniline, [3-(Phenylamino)propyl]trimethoxysilane
Linear Formula:
C6H5NH(CH2)3Si(OCH3)3
CAS Number:
Molecular Weight:
255.39
Products
Shipping
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing