Skip to Content
Merck
N-[(1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide certified reference material, pharmaceutical secondary standard

N-[(1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide

N-[(1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide certified reference material, pharmaceutical secondary standard
Empirical Formula (Hill Notation):
C17H22N4O
CAS Number:
Molecular Weight:
298.38

Products

Brand
Formula Weight
Quality Segment
Application
Package Size
API Family
Special Grade