Skip to Content
Merck
N-(1-Pyrenyl)maleimide

N-(1-Pyrenyl)maleimide

N-(1-Pyrenyl)maleimide
Synonym(s):
1-(1-Pyrenyl)-1H-pyrrole-2,5-dione
Empirical Formula (Hill Notation):
C20H11NO2
CAS Number:
Molecular Weight:
297.31
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing