Skip to Content
Merck
Monoclonal Anti-Maltose Binding Protein−Peroxidase antibody produced in mouse clone MBP-17, purified immunoglobulin, lyophilized powder

Monoclonal Anti-Maltose Binding Protein−Peroxidase antibody produced in mouse

Monoclonal Anti-Maltose Binding Protein−Peroxidase antibody produced in mouse clone MBP-17, purified immunoglobulin, lyophilized powder
Synonym(s):
Monoclonal Anti-Maltose Binding Protein, Anti-MBP

Products

Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Package Size
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing