![Micro particles based on melamine resin, rhodamine B-marked size: 5 μm](/deepweb/assets/sigmaaldrich/product/images/316/273/fe113794-e14f-4449-a4de-2b656a3342ff/640/fe113794-e14f-4449-a4de-2b656a3342ff.jpg)
Micro particles based on melamine resin, rhodamine B-marked
![Micro particles based on melamine resin, rhodamine B-marked size: 5 μm](/deepweb/assets/sigmaaldrich/product/images/316/273/fe113794-e14f-4449-a4de-2b656a3342ff/640/fe113794-e14f-4449-a4de-2b656a3342ff.jpg)
Synonym(s):
Melamine-formaldehyde resin
Products
Product Category
Brand
Physical Form
Format
Quality Segment
Application
Particle Size (µm)
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing