![Methylrosanilinium for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/131/023/133aac8c-c308-4599-8bd3-a96fe7ba1373/640/133aac8c-c308-4599-8bd3-a96fe7ba1373.png)
Methylrosanilinium for system suitability
![Methylrosanilinium for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/131/023/133aac8c-c308-4599-8bd3-a96fe7ba1373/640/133aac8c-c308-4599-8bd3-a96fe7ba1373.png)
Synonym(s):
Crystal Violet, Basic Violet 3, Gentian Violet, Hexamethylpararosaniline chloride, Methyl Violet 10B
Linear Formula:
C25H30N3Cl
CAS Number:
Molecular Weight:
407.98
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing