Skip to Content
Merck
Methyl acetopyruvate 97%

Methyl acetopyruvate

Methyl acetopyruvate 97%
Synonym(s):
1-Methoxy-1,2,4-pentanetrione, Methyl β-acetylpyruvate, Methyl 2,4-dioxopentanoate, Methyl 2,4-dioxovalerate, Methyl 3-acetylpyruvate
Empirical Formula (Hill Notation):
C6H8O4
CAS Number:
Molecular Weight:
144.13
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing