Skip to Content
Merck
Metalaxyl-(phenyl-13C6) 99 atom % 13C, 98% (CP)

Metalaxyl-(phenyl-13C6)

Metalaxyl-(phenyl-13C6) 99 atom % 13C, 98% (CP)
Empirical Formula (Hill Notation):
13C6C9H21NO4
CAS Number:
Molecular Weight:
285.29

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing