Skip to Content
Merck
Mepanipyrim PESTANAL®, analytical standard

Mepanipyrim

Mepanipyrim PESTANAL®, analytical standard
Synonym(s):
4-Methyl-N-phenyl-6-(1-propynyl)-2-pyrimidinamine
Empirical Formula (Hill Notation):
C14H13N3
CAS Number:
Molecular Weight:
223.27
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing