Skip to Content
Merck
Meldonium dihydrate European Pharmacopoeia (EP) Reference Standard

Meldonium dihydrate

Meldonium dihydrate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
MET 88, Quaterin
Empirical Formula (Hill Notation):
C6H14N2O2 · 2H2O
CAS Number:
Molecular Weight:
182.22

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2