![Madecassoside from Centella asiatica ≥95% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/153/007/881ae103-563f-4cf4-98e0-2fb225c934c2/640/881ae103-563f-4cf4-98e0-2fb225c934c2.png)
Madecassoside from Centella asiatica
![Madecassoside from Centella asiatica ≥95% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/153/007/881ae103-563f-4cf4-98e0-2fb225c934c2/640/881ae103-563f-4cf4-98e0-2fb225c934c2.png)
Synonym(s):
Madecassoside, Asiaticoside A
Empirical Formula (Hill Notation):
C48H78O20
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Research Area
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing